Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat RAGE Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
$200.50 - $549.50
Specifications
Antigen | RAGE |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL |
Applications | Western Blot, Blockade of Receptor-ligand Interaction, ELISA |
Classification | Polyclonal |
Description
AGER Polyclonal specifically detects AGER in Rat samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair).Specifications
RAGE | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL | |
Polyclonal | |
Goat | |
Rat | |
advanced glycosylation end product-specific receptor, AGER, RAGE isoform delta, RAGE isoform sRAGE-delta, Receptor for advanced glycosylation end products, receptor for advanced glycosylation end-products, SCARJ1 | |
Ager | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Blockade of Receptor-ligand Interaction, ELISA | |
Unconjugated | |
RUO | |
Q63495 | |
5891 | |
Mouse myeloma cell line NS0-derived recombinant rat RAGE Gln24-Ala342 Accession # Q63495 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects rat RAGE in ELISAs and Western blots. In sandwich immunoassays, less than 0.1% cross-reactivity with recombinant human RAGE and recombinant canine RAGE is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title