Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat TSH beta Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $538.00
Specifications
Antigen | TSH beta |
---|---|
Clone | 442506 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
TSH beta Monoclonal specifically detects TSH beta in Rat samples. It is validated for Western Blot.Specifications
TSH beta | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Rat | |
P04652 | |
7252 | |
E. coli-derived recombinant rat TSH beta Phe21-Gly138 Accession # P04652 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects rat TSH beta in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows approximately 5% cross-reactivity with recombinant human TSH beta . |
442506 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, Thyrotropin beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSHB, TSH-B, TSH-BETA | |
TSHB | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title