Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat UNC5H2/UNC5B Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
$188.28 - $686.23
Specifications
| Antigen | UNC5H2/UNC5B |
|---|---|
| Clone | 187305 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
Description
UNC5H2/UNC5B Monoclonal specifically detects UNC5H2/UNC5B in Rat samples. It is validated for Western Blot.Specifications
| UNC5H2/UNC5B | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Rat | |
| O08722 | |
| 219699 | |
| Mouse myeloma cell line NS0-derived recombinant rat UNC5H2 Gly27-Asp373 Accession # O08722 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects rat UNC5H2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant rat UNC5H1 or recombinant human UNC5H4 is observed. |
| 187305 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| p53RDL1, p53-regulated receptor for death and life protein 1, Protein unc-5 homolog 2, Protein unc-5 homolog B, transmembrane receptor Unc5H2, unc5 (C.elegans homolog) b, unc-5 homolog 2, unc-5 homolog B (C. elegans), UNC5B, UNC5H2netrin receptor UNC5B | |
| UNC5B | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title