Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rattin Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB018153 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018153 0.025 mL
NB300246 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018153 Supplier Novus Biologicals Supplier No. NB300246SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

Rattin Polyclonal specifically detects Rattin in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.

Specifications

Antigen Rattin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Immunohistochemistry-Paraffin
Formulation PBS with 0.1% Sodium Azide
Gene Alias Humanin-like in rat, rattin humanin-like
Host Species Rabbit
Immunogen A synthetic peptide mapping to residues 20-35 [VKRLESPNKTRRPYGA] of the Rattin protein.
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Test Specificity Reacts with residues 20-35 [VKRLESPNKTRRPYGA] of the Rattin peptide, which is not in the conserved region to Humanin.
Target Species Human, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.