Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rattin Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB344588 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344588 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344588 Supplier Novus Biologicals Supplier No. NB300246MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rattin Polyclonal antibody specifically detects Rattin in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)

Specifications

Antigen Rattin
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Humanin-like in rat, rattin humanin-like
Host Species Rabbit
Immunogen A synthetic peptide mapping to residues 20-35 [VKRLESPNKTRRPYGA] of the Rattin protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.