Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Raw 264.7 (mouse macrophage, Abelson murine leukemia virus transformed) Whole cell lysate, Denatured; Abnova
Click to view available options
Quantity:
200 μg
Description
Western Blotting
Specifications
Specifications
Tissue | Macrophage |
For Use With (Application) | Western Blot |
Description | Whole cell lysate (denatured) |
Lysis Buffer | Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. |
Preparation Method | Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). The lysate was mixed in 5X Sample Buffer to become final 1X Sample Buffer as Storage Buffer. The lysate was heated at 95°C for 5 min, and cooled rapidly. |
Quality Control Testing | 12.5% SDS-PAGE Stained with Coomassie Blue. |
Quantity | 200 μg |
Storage Buffer | In ready-to-use 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue). |
Host Species | Mouse |
Content And Storage | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction