Learn More
Description
Specifications
Specifications
| Antigen | RAX |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | MCOP3, retina and anterior neural fold homeobox, retinal homeobox protein Rx, RXRetina and anterior neural fold homeobox protein |
| Gene Symbols | RAX |
| Host Species | Rabbit |
| Immunogen | A portion of amino acids 300-350 of human Rax was used as the immunogen. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
