Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB1 Antibody (172C1094), Alexa Fluor™ 700 - (cleaved), Novus Biologicals™
SDP

Catalog No. NB10056598Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056598Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056598Z Supplier Novus Biologicals Supplier No. NB10056598AF700

Mouse Monoclonal Antibody

RB1 Monoclonal specifically detects RB1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.

Specifications

Antigen cleaved Retinoblastoma 1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown
Classification Monoclonal
Clone 172C1094
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
Gene Symbols RB1
Host Species Mouse
Immunogen A synthetic peptide corresponding to a region of the caspase-3-induced cleavage site of human Rb was used as immunogen.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5925
Test Specificity The antibody recognizes cleaved Rb (DRb-p70). The antibody does not recognize full-length Rb. In camptothecin treated HL60, an ∼68 kDa band is observed. DRb-p70 exists in vivo as an N-terminally truncated form of Rb (Liu et al.). DRb-p70 is thought to be the product of alternative translation and is expressed in primary myeloid cells in fetal liver, bone marrow and spleen. DRb-p70 is also specifically expressed in tumor-associated macrophages (TAMs). TAMs are an important targets of cancer therapy. DRb-p70 is a marker of TAMs, and maybe useful for identifying tumors with macrophage infiltration (Liu et al, 2005).
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.