Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB1 Antibody (1F8), Novus Biologicals™
SDP

Catalog No. NB253398200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB253398200 200 μg
NB25339820 20 μg
NB253398100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB253398200 Supplier Novus Biologicals Supplier No. NBP253398200UG

Mouse Monoclonal Antibody

RB1 Monoclonal antibody specifically detects RB1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen RB1
Applications Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1F8
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Immunoprecipitation 0.5 - 1 ug, Immunohistochemistry-Paraffin 1 - 2 ug/ml
Formulation 10mM PBS with 0.05% BSA with 0.05% Sodium Azide
Gene Alias OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
Gene Symbols RB1
Host Species Mouse
Immunogen Recombinant human Rb1 protein fragment of 283 amino acid residues (exact sequence is proprietary)
Molecular Weight of Antigen 105 kDa
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5925
Target Species Human, Mouse
Content And Storage Store at 4C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.