Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056888 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
0.2 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056888 0.2 mL
NB10056888T 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056888 Supplier Novus Biologicals Supplier No. NB10056888
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RB1 Polyclonal specifically detects RB1 in Canine, Equine, Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen RB1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:200-1:1000
Formulation PBS containing 0.2% gelatin with 0.05% Sodium Azide
Gene Accession No. P06400
Gene Alias OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
Gene Symbols RB1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 251-268 (RTPRRGQNRSARIAKQLE) of human Rb was used as immunogen
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5925
Test Specificity In Jurkat, a 110 kDa band is observed.
Target Species Canine, Mouse, Equine, Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.