Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB1 Antibody (RB1/2313R), Biotin, Novus Biologicals™
SDP

Catalog No. NB389208 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389208 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389208 Supplier Novus Biologicals Supplier No. NBP308940B
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RB1 Monoclonal antibody specifically detects RB1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen RB1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone RB1/2313R
Conjugate Biotin
Formulation PBS
Gene Alias OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
Host Species Rabbit
Immunogen Recombinant human full-length RB1 protein (Uniprot: P06400)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5925
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.