Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB1, p Ser807 Antibody, Novus Biologicals™
SDP

Catalog No. NB017880 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.05 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017880 0.025 mL
NB10082177 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017880 Supplier Novus Biologicals Supplier No. NB100821770.025ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RB1 (p Ser807) Polyclonal specifically detects RB1 (p Ser807) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen RB1 (p Ser807)
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100
Formulation PBS [pH 7.4] 150mM NaCl and 50% glycerol (without Mg2+ and Ca2+) with 0.02% Sodium Azide
Gene Accession No. P06400
Gene Alias OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein
Gene Symbols RB1
Host Species Rabbit
Immunogen The antiserum was produced against synthesized phosphopeptide derived from human Rb around the phosphorylation site of serine 807 (Y-I-SP-P-L).
Molecular Weight of Antigen 110 kDa
Purification Method Affinity Purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5925
Test Specificity Rb (specific to Phospho-Ser807) detects endogenous levels of Rb only when phosphorylated at serine 807.
Target Species Human, Mouse, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.