Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB1, p Ser807 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$280.00 - $438.00
Specifications
Antigen | RB1 (p Ser807) |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017880
![]() |
Novus Biologicals
NB100821770.025ML |
0.025 mL |
Each for $280.00
|
|
|||||
NB10082177
![]() |
Novus Biologicals
NB10082177 |
0.05 mL |
Each for $438.00
|
|
|||||
Description
RB1 Polyclonal specifically detects RB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RB1 (p Ser807) | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
Polyclonal | |
Rabbit | |
Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
P06400 | |
5925 | |
IgG | |
Affinity Purified | |
110 kDa |
1.0 mg/mL | |
Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat | |
OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
RB1 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title