Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB545 Mouse Anti-T-bet, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569758
Description
T-bet Monoclonal antibody specifically detects T-bet in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)
Specifications
T-bet | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human, Mouse | |
Liquid |
Flow Cytometry | |
4B10 | |
RB545 | |
T-box expressed in T cells; TBX21; T-box 21; TBLYM | |
Mouse T-bet Recombinant Protein | |
25 μg | |
30009, 57765 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction