Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB545 Mouse Anti-T-bet, BD Horizon™
Mouse Monoclonal Antibody
$182.00 - $426.00
Specifications
Antigen | T-bet |
---|---|
Clone | 4B10 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
T-bet Monoclonal antibody specifically detects T-bet in Human|Mouse samples. It is validated for Intracellular staining (flow cytometry)Specifications
T-bet | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
30009, 57765 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
4B10 | |
Flow Cytometry | |
RB545 | |
Mouse | |
Human, Mouse | |
T-box expressed in T cells; TBX21; T-box 21; TBLYM | |
Mouse T-bet Recombinant Protein | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title