Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Hamster Anti-Mouse CD3e, BD Horizon™
Hamster Monoclonal Antibody
$104.00 - $340.00
Specifications
Antigen | CD3e |
---|---|
Clone | 145-2C11 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD3e Monoclonal antibody specifically detects CD3e in Mouse samples. It is validated for Flow cytometrySpecifications
CD3e | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
12501 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
145-2C11 | |
Flow Cytometry | |
RB705 | |
Hamster | |
Mouse | |
CD3; CD3 epsilon; Cd3e; CD3ε; T3e | |
H-2Kb specific cytotoxic T lymphocyte clone BM10-37 | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title