Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD123 (IL-3 Receptor α), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570252
Description
CD123 (IL-3 Receptor α) Monoclonal antibody specifically detects CD123 (IL-3 Receptor α) in Human samples. It is validated for Flow cytometry
Specifications
| CD123 (IL-3 Receptor α) | |
| Monoclonal | |
| RB705 | |
| IL3RA; IL-3RA; IL-3Rα; IL-3R-alpha; Interleukin-3 receptor subunit alpha | |
| Human IL-3Ra-transfected cells | |
| 25 Tests | |
| 3563 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
| IgG2a κ |
| Flow Cytometry | |
| 7G3 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction