Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD123 (IL-3 Receptor α), BD Horizon™
Mouse Monoclonal Antibody
$168.00 - $412.00
Specifications
| Antigen | CD123 (IL-3 Receptor α) |
|---|---|
| Clone | 7G3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | RB705 |
Description
CD123 (IL-3 Receptor α) Monoclonal antibody specifically detects CD123 (IL-3 Receptor α) in Human samples. It is validated for Flow cytometrySpecifications
| CD123 (IL-3 Receptor α) | |
| Flow Cytometry | |
| RB705 | |
| Mouse | |
| Human | |
| IL3RA; IL-3RA; IL-3Rα; IL-3R-alpha; Interleukin-3 receptor subunit alpha | |
| Human IL-3Ra-transfected cells | |
| Affinity Chromatography |
| 7G3 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 3563 | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title