Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD195 (CCR5), BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570657
Description
CD195 (CCR5) Monoclonal antibody specifically detects CD195 (CCR5) in Human samples. It is validated for Flow cytometry
Specifications
CD195 (CCR5) | |
Monoclonal | |
RB705 | |
CCR5; C-C chemokine receptor type 5; CC-CKR-5; CKR5; CHEMR13 | |
Human CCR5 Transfected Cell Line | |
25 Tests | |
1234 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Flow Cytometry | |
2D7/CCR5 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction