Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD195 (CCR5), BD Horizon™
Mouse Monoclonal Antibody
$160.00 - $406.00
Specifications
| Antigen | CD195 (CCR5) |
|---|---|
| Clone | 2D7/CCR5 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | RB705 |
Description
CD195 (CCR5) Monoclonal antibody specifically detects CD195 (CCR5) in Human samples. It is validated for Flow cytometrySpecifications
| CD195 (CCR5) | |
| Flow Cytometry | |
| RB705 | |
| Mouse | |
| Human | |
| CCR5; C-C chemokine receptor type 5; CC-CKR-5; CKR5; CHEMR13 | |
| Human CCR5 Transfected Cell Line | |
| Affinity Chromatography |
| 2D7/CCR5 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 1234 | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title