Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD33, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 570727
Description
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow cytometry
Specifications
CD33 | |
Monoclonal | |
RB705 | |
Siglec-3; SIGLEC3; Sialic acid-binding Ig-like lectin 3; p67; gp67; My9 | |
Acute Myeloid Leukemia Blasts | |
25 Tests | |
945 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 κ |
Flow Cytometry | |
WM53 (also known as WM-53) | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction