Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Mouse Anti-Human CD33, BD Horizon™
Mouse Monoclonal Antibody
$192.00 - $412.00
Specifications
| Antigen | CD33 |
|---|---|
| Clone | WM53 (also known as WM-53) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | RB705 |
Description
CD33 Monoclonal antibody specifically detects CD33 in Human samples. It is validated for Flow cytometrySpecifications
| CD33 | |
| Flow Cytometry | |
| RB705 | |
| Mouse | |
| Human | |
| Siglec-3; SIGLEC3; Sialic acid-binding Ig-like lectin 3; p67; gp67; My9 | |
| Acute Myeloid Leukemia Blasts | |
| Affinity Chromatography |
| WM53 (also known as WM-53) | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 945 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title