Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB705 Rat Anti-Mouse Ly-6A/E (Sca-1), BD Horizon™
Rat Monoclonal Antibody
$120.00 - $298.00
Specifications
Antigen | Ly-6A/E (Sca-1) |
---|---|
Clone | D7 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
Ly-6A/E (Sca-1) Monoclonal antibody specifically detects Ly-6A/E (Sca-1) in Mouse samples. It is validated for Flow cytometrySpecifications
Ly-6A/E (Sca-1) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
110454 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
D7 | |
Flow Cytometry | |
RB705 | |
Rat | |
Mouse | |
Ly-6A/E; Lymphocyte antigen 6A-2/6E-1; Ly-6A.2/Ly-6E.1; Sca-1; TAP | |
IL-2-dependent mouse T-cell line CTL-L | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title