Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB780 Rat Anti-Mouse CD25 (IL-2 Receptor α), BD Horizon™
Rat Monoclonal Antibody
$98.00 - $236.00
Specifications
Antigen | CD25 (IL-2 Receptor α) |
---|---|
Clone | PC61 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD25 (IL-2 Receptor α) Monoclonal antibody specifically detects CD25 (IL-2 Receptor α) in Mouse samples. It is validated for Flow cytometrySpecifications
CD25 (IL-2 Receptor α) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
16184 | |
IgG1 λ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
PC61 | |
Flow Cytometry | |
RB780 | |
Rat | |
Mouse | |
Interleukin-2 receptor alpha chain; IL-2RA; IL-2Rα; Il2ra; IL-2R p55 | |
IL-2-dependent cytolytic mouse T-cell clone B6.1 | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title