Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RbAp46 Antibody, Novus Biologicals™
SDP

Catalog No. NB1203535 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1203535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1203535 Supplier Novus Biologicals Supplier No. NB1203535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

RbAp46 Polyclonal specifically detects RbAp46 in Human, Mouse, Herring samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RbAp46
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml, Immunohistochemistry 1.0 ug/ml, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Formulation PBS containing 1 mg/ml BSA with 0.05% Sodium Azide
Gene Alias G1/S transition control protein-binding protein RbAp46, Histone acetyltransferase type B subunit 2, histone-binding protein RBBP7, MGC138867, Nucleosome-remodeling factor subunit RBAP46, RbAp46, RBBP-7, retinoblastoma binding protein 7, Retinoblastoma-binding protein 7MGC138868, Retinoblastoma-binding protein p46, retinoblastoma-binding protein RbAp46
Gene Symbols RBBP7
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues M(1) A S K E M F E D T V E E(13) C of human RbAp46.
Molecular Weight of Antigen 46 kDa
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Growth and Development, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5931
Test Specificity NB120-3535 detects retinoblastoma protein (Rb) associated protein 46 (RbAp46) from human cells.
Target Species Human, Mouse, Herring
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.