Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RbAp46 Antibody (OTI5A4), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N273822J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273822J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273822J549 Supplier Novus Biologicals Supplier No. NBP273822JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RbAp46 Monoclonal specifically detects RbAp46 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen RbAp46
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI5A4
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias G1/S transition control protein-binding protein RbAp46, Histone acetyltransferase type B subunit 2, histone-binding protein RBBP7, MGC138867, Nucleosome-remodeling factor subunit RBAP46, RbAp46, RBBP-7, retinoblastoma binding protein 7, Retinoblastoma-binding protein 7MGC138868, Retinoblastoma-binding protein p46, retinoblastoma-binding protein RbAp46
Gene Symbols RBBP7
Host Species Mouse
Immunogen Full length recombinant protein of human RbAp46 (NP_002884) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Chromatin Research, Growth and Development, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5931.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.