Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP 4/7 Antibody (968721), R&D Systems™
SDP

Catalog No. MAB9457SP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9457SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB9457SP Supplier Novus Biologicals Supplier No. MAB9457SP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBBP 4/7 Monoclonal specifically detects RBBP 4/7 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen RBBP 4/7
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 968721
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry
Formulation PBS with No Preservative
Host Species Mouse
Immunogen E. coli-derived recombinant human RBBP-4
Purification Method Protein A or G purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.