Learn More
Description
Specifications
Specifications
| Antigen | RBBP4/RbAp48 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 15G12 |
| Concentration | 1.0mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunoprecipitation 0.2-1 ug/ml, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Gene Alias | CAF-1 subunit C, CAF-I p48, Chromatin assembly factor 1 subunit C, Chromatin assembly factor I p48 subunit, chromatin assembly factor/CAF-1 p48 subunit, histone-binding protein RBBP4, MSI1 protein homolog, Nucleosome-remodeling factor subunit RBAP48, NURF55, RbAp48, RBBP-4, retinoblastoma binding protein 4, Retinoblastoma-binding protein 4CAF-I 48 kDa subunit, Retinoblastoma-binding protein p48 |
| Gene Symbols | RBBP4 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
