Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP6 Antibody, Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP149542Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149542Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149542Z Supplier Novus Biologicals Supplier No. NBP149542AF700
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBBP6 Polyclonal specifically detects RBBP6 in Human samples. It is validated for Western Blot.

Specifications

Antigen RBBP6
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. Q7Z6E9
Gene Alias E3 ubiquitin-protein ligase RBBP6, EC 6.3.2.-, MY038, P2PR, P2P-R, p53-associated cellular protein of testis, PACTDKFZp761B2423, Proliferation potential-related protein, Protein P2P-R, RB-binding Q-protein 1, RBQ1, RBQ-1, retinoblastoma binding protein 6, Retinoblastoma-binding protein 6DKFZp686P0638, Retinoblastoma-binding Q protein 1, SNAMA
Gene Symbols RBBP6
Host Species Rabbit
Immunogen A synthetic peptide made to an internal portion of the human RBBP6 protein (between residues 20-70) [UniProt Q7Z6E9]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 5930
Test Specificity Human. Predicted to react with mouse based on 100% sequence homology.
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.