Learn More
RbBP7 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc IHC00160

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Dog, Horse, Guinea pig_10141, Pig, Panda, Orangutan, Monkey, Chimpanzee, Naked mole r
This protein is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that binds directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein is found in many histone deacetylase complexes, including mSin3 co-repressor complex. It is also present in protein complexes involved in chromatin assembly. This protein can interact with BRCA1 tumor-suppressor gene and may have a role in the regulation of cell proliferation and differentiation.Specifications
RbBP7 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q16576, Q60973 | |
Rbbp7 | |
Between 375 and C-term | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry | |
Unconjugated | |
RBBP7 | |
Histone-binding protein RBBP7, Retinoblastoma-binding protein 7, RBBP-7, Retinoblastoma-binding protein p46, Histone acetyltransferase type B subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
245688, 5931 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.