Learn More
RbBP7 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | RbBP7 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1600
![]() |
Bethyl Laboratories, Inc
IHC00160T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1599
![]() |
Bethyl Laboratories, Inc
IHC00160 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Dog, Horse, Guinea pig_10141, Pig, Panda, Orangutan, Monkey, Chimpanzee, Naked mole r
This protein is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that binds directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein is found in many histone deacetylase complexes, including mSin3 co-repressor complex. It is also present in protein complexes involved in chromatin assembly. This protein can interact with BRCA1 tumor-suppressor gene and may have a role in the regulation of cell proliferation and differentiation.Specifications
RbBP7 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Histone-binding protein RBBP7, Retinoblastoma-binding protein 7, RBBP-7, Retinoblastoma-binding protein p46, Histone acetyltransferase type B subunit | |
Between 375 and C-term | |
Primary | |
4° C | |
RBBP7 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q16576, Q60973 | |
245688, 5931 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.