Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP9 Antibody (OTI4G3), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N273828J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N273828J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N273828J549 Supplier Novus Biologicals Supplier No. NBP273828JF549

Mouse Monoclonal Antibody

RBBP9 Monoclonal specifically detects RBBP9 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RBBP9
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4G3
Conjugate Janelia Fluor 549
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias B5T-overexpressed gene protein, Bog, BOGB5T overexpressed gene protein, EC 3.-, Protein BOG, putative hydrolase RBBP9, RBBP10, RBBP-10, RBBP-9, retinoblastoma binding protein 9, Retinoblastoma-binding protein 10, Retinoblastoma-binding protein 9MGC9236, retinoma-binding protein 9
Gene Symbols RBBP9
Host Species Mouse
Immunogen Full length recombinant protein of human RBBP9 (NP_006597) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10741.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.