Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBCK1 Antibody, DyLight 350, Novus Biologicals™
SDP

Catalog No. NB052346 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052346 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052346 Supplier Novus Biologicals Supplier No. NBP298085UV
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBCK1 Polyclonal specifically detects RBCK1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RBCK1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias C20orf18, EC 6.3.2.-, HBV associated factor 4, HBV-associated factor 4, Heme-oxidized IRP2 ubiquitin ligase 1, Hepatitis B virus X-associated protein 4, HOIL-1, RanBP-type and C3HC4-type zinc finger containing 1, ranBP-type and C3HC4-type zinc finger-containing protein 1, RBCC protein interacting with PKC1, RBCK2, RNF54RING finger protein 54, UBCE7IP3HOIL1, ubiquitin conjugating enzyme 7 interacting protein 3, Ubiquitin-conjugating enzyme 7-interacting protein 3, XAP3, XAP4chromosome 20 open reading frame 18, ZRANB4
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RBCK1 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 10616
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.