Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBED1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP179821

 View more versions of this product

Catalog No. NBP179821


Only null left
Add to Cart

Description

Description

RBED1 Polyclonal specifically detects RBED1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RBED1
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
C330008I15Rik, ELMO domain-containing protein 3, ELMO/CED-12 domain containing 3, FLJ21977, FLJ35601, liver-specific organic anion transporter 3TM12, LST3, MGC111036, organic anion transporter LST-3b, RBED1, RBM29, RNA binding motif and ELMO domain 1, RNA binding motif and ELMO/CED-12 domain 1, RNA binding motif protein 29, RNA-binding motif and ELMO domain-containing protein 1, RNA-binding motif protein 29, RNA-binding protein 29
Rabbit
44 kDa
100 μL
Apoptosis
84173
Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Guinea Pig, Rabbit
IgG
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
NP_115589
ELMOD3
Synthetic peptide directed towards the C terminal of human ELMOD3. Peptide sequence FRLSRHHIQQFPFCLMSVNITHIAIQALREECLSRECNRQQKVIPVVNSF.
Affinity purified
RUO
Primary
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.