Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBFOX3/NeuN Antibody (1B7), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB192693594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB192693594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB192693594 Supplier Novus Biologicals Supplier No. NBP192693DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBFOX3/NeuN Monoclonal specifically detects RBFOX3/NeuN in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).

Specifications

Antigen RBFOX3/NeuN
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1B7
Conjugate DyLight 594
Dilution Western Blot 1:5000 - 1:10000, Flow Cytometry 1:10, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:500 - 1:1000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:400, Flow (Intracellular)
Formulation 50 mM Sodium Borate with 0.05% Sodium Azide
Gene Alias FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, NeuN flow cytometry, neuronal nuclei, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3
Gene Symbols RBFOX3
Host Species Mouse
Immunogen N-terminal 99 amino acids of human FOX3 as expressed in and purified from E. coli
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 146713
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.