Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBFOX3/NeuN Antibody (NeuN/288R), HRP, Novus Biologicals™
SDP

Catalog No. NB178019 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB178019 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB178019 Supplier Novus Biologicals Supplier No. NBP314002H
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

RBFOX3/NeuN Monoclonal antibody specifically detects RBFOX3/NeuN in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen RBFOX3/NeuN
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone NeuN/288R
Conjugate HRP
Formulation PBS
Gene Alias FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuN antigen, neuronal nuclei, neuronal nuclei antigen, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3, RNA binding protein, fox-1 homolog 3
Host Species Rabbit
Immunogen Recombinant full-length human RBFOX3/NeuN protein (Uniprot: A6NFN3)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 146713
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.