Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM14 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB292233 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292233 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292233 Supplier Novus Biologicals Supplier No. NB1002884APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBM14 Polyclonal specifically detects RBM14 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen RBM14
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias COAAMGC31756, Paraspeckle protein 2, PSP2DKFZp779J0927, RNA binding motif protein 14, RNA-binding motif protein 14, RNA-binding protein 14, RRM-containing coactivator activator/modulator, SIPMGC15912, Synaptotagmin-interacting protein, SYT-interacting protein, SYTIP1, TMEM137, transmembrane protein 137
Host Species Rabbit
Immunogen Maps to a region between residue 550 and 600 of human CoActivator Activator (RNA Binding Motif 14) using the numbering given in entry NP_006319.1 (GeneID 10432 ).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, DNA Repair
Primary or Secondary Primary
Gene ID (Entrez) 10432
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.