Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM15 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB056566 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056566 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056566 Supplier Novus Biologicals Supplier No. NBP298922AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBM15 Polyclonal specifically detects RBM15 in Human samples. It is validated for Western Blot.

Specifications

Antigen RBM15
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias FLJ12479, one twenty two protein, one twenty-two, One-twenty two protein 1, putative RNA-binding protein 15, RNA binding motif protein 15, RNA-binding motif protein 15, SPEN
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human RBM15.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64783
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.