Learn More
RBM15 Polyclonal Antibody, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt.
Specifications
Specifications
| Antigen | RBM15 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.1% BSA and 0.09% sodium azide |
| Gene | RBM15 |
| Gene Accession No. | Q96T37 |
| Gene Alias | OTT, OTT1, SPEN, RNA-binding motif protein 15, one twenty two protein, one twenty-two, one-twenty two protein 1, putative RNA-binding protein 15 |
| Gene Symbols | RBM15 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.