Learn More
RBM15B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | RBM15B |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-3796
![]() ![]() |
Proteintech Group Inc
222491AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This RBM15B antibody was produced by immunizing animals with the C-terminus of RBM15B, which is homologous to a short form of RBM15B (563 aa, ∽70 kDa), thus this antibody can recognize full length of RBM15B (∽100 kDa) and splice-one (70 kDa).
May function in the regulation of alternative or illicit splicing.Specifications
RBM15B | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q6PHZ5, Q8NDT2 | |
109095, 29890, 315988 | |
RBM15B Fusion Protein Ag17744 | |
Primary | |
-20°C | |
RBM15B |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HUMAGCGB, HuOTT3, One twenty two protein 3, OTT3, RBM15B, RNA binding motif protein 15B | |
Rbm15b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.