Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM18 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1804720 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1804720 20 μL
NBP180471 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1804720 Supplier Novus Biologicals Supplier No. NBP18047120UL

Rabbit Polyclonal Antibody

RBM18 Polyclonal specifically detects RBM18 in Human samples. It is validated for Western Blot.

Specifications

Antigen RBM18
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. NP_149108
Gene Alias MGC2734, RNA binding motif protein 18, RNA-binding motif protein 18
Gene Symbols RBM18
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RBM18. Peptide sequence VRWAHAQVKRYDHNKNDKILPISLEPSSSTEPTQSNLSVTAKIKAIEAKL.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92400
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.