Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM19 Antibody, Novus Biologicals™
SDP

Catalog No. NB432818 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB432818 25 μL
NBP184135 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB432818 Supplier Novus Biologicals Supplier No. NBP18413525UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

RBM19 Polyclonal specifically detects RBM19 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen RBM19
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias KIAA0682DKFZp586F1023, NPO, probable RNA-binding protein 19, RNA binding motif protein 19, RNA-binding motif protein 19
Gene Symbols RBM19
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:PKNTTKSWQGRILGENEEEEDLAESGRLFVRNLPYTSTEEDLEKLFSKYGPLSELHYPIDSLTKKPKGFAFITFMFPEHAVKAYSEVDGQVFQGRMLHVLPSTIKKEASE
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9904
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.