Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM20 Antibody, Novus Biologicals™
SDP

Catalog No. NBP227509 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227509 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP227509 Supplier Novus Biologicals Supplier No. NBP227509
Add to Cart
Add to Cart

Goat Polyclonal Antibody

RBM20 Polyclonal specifically detects RBM20 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen RBM20
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3-1 ug/ml, Peptide ELISA Detection limit 1:164000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_001127835.1
Gene Alias RNA binding motif protein 20
Gene Symbols RBM20
Host Species Goat
Immunogen Peptide with sequence C-QLDKAELDERP corresponding to internal region according to NP_001127835.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282996
Test Specificity Reacts with Mouse. Expected from sequence similarity: Human, Rat and Cow.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.