Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM22 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB290343 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290343 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290343 Supplier Novus Biologicals Supplier No. NBP222275PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBM22 Polyclonal antibody specifically detects RBM22 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen RBM22
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias Cwc2, FLJ10290, fSAP47, functional spliceosome-associated protein 47, pre-mRNA-splicing factor RBM22, RNA binding motif protein 22, ZC3H16RNA-binding motif protein 22, Zinc finger CCCH domain-containing protein 16
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 175 and 225 of human RNA Binding Motif Protein 22 using the numbering given in entry NP_060517.1 (GeneID 55696).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55696
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.