Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM25/RED120 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140618
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-9165 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-9165 Supplier Bethyl Laboratories, Inc Supplier No. A301068AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

RNA-binding protein that acts as a regulator of alternative pre-mRNA splicing. Involved in apoptotic cell death through the regulation of the apoptotic factor BCL2L1 isoform expression. Modulates the ratio of proapoptotic BCL2L1 isoform S to antiapoptotic BCL2L1 isoform L mRNA expression. When overexpressed, stimulates proapoptotic BCL2L1 isoform S 5′-splice site (5′-ss) selection, whereas its depletion caused the accumulation of antiapoptotic BCL2L1 isoform L. Promotes BCL2L1 isoform S 5′-ss usage through the 5′-CGGGCA-3′ RNA sequence. Its association with LUC7L3 promotes U1 snRNP binding to a weak 5′ ss in a 5′-CGGGCA-3′-dependent manner. Binds to the exonic splicing enhancer 5′-CGGGCA-3′ RNA sequence located within exon 2 of the BCL2L1 pre-mRNA. Also involved in the generation of an abnormal and truncated splice form of SCN5A in heart failure.

Specifications

Antigen RBM25/RED120
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene RBM25
Gene Accession No. B2RY56, P49756
Gene Alias arg/Glu/Asp-rich protein of 120 kDa, Arg/Glu/Asp-rich protein, 120 kDa, fSAP94, functional spliceosome-associated protein 94, NET52, Protein S164, RED120, RNA-binding motif protein 25, RNA-binding region (RNP1, RRM) containing 7, RNA-binding region-containing protein 7, RNPC7, S164, Snu71, U1 small nuclear ribonucleoprotein 1SNRP homolog
Gene Symbols Rbm25
Host Species Rabbit
Immunogen Between 475 and 525
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58517, 67039
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.