Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM26 Antibody, Novus Biologicals™
SDP

Catalog No. NB10060409 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10060409 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10060409 Supplier Novus Biologicals Supplier No. NB10060409
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBM26 Polyclonal specifically detects RBM26 in Human samples. It is validated for Immunoprecipitation, Western Blot.

Specifications

Antigen RBM26
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_071401.3
Gene Alias acidic rich RS domain containing 2, ARRS2, C13orf10, chromosome 13 open reading frame 10, CTCL tumor antigen se70-2, cutaneous T-cell lymphoma tumor antigen se70-2, FLJ20957, MGC133295, MGC133296, PRO1777, RNA binding motif protein 26, RNA-binding motif protein 26, RNA-binding protein 26, RP11-255E21.1, SE70-2, ZC3H17
Gene Symbols RBM26
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 450 and 500 of human RNA binding motif protein 26 using the numbering given in entry NP_071401.3 (GeneID 64062 ).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64062
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.