Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM33 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB290331 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290331 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290331 Supplier Novus Biologicals Supplier No. NBP222280PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBM33 Polyclonal antibody specifically detects RBM33 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative)

Specifications

Antigen RBM33
Applications Western Blot
Classification Polyclonal
Conjugate PE
Dilution Western Blot, Immunoprecipitation (Negative)
Formulation PBS
Gene Alias DKFZp434D1319, MGC20460, proline-rich protein 8, RNA binding motif protein 33, RNA-binding motif protein 33, RNA-binding protein 33
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 500 and 550 of human RNA Binding Motif Protein 33 using the numbering given in entry NP_444271.2 (GeneID 155435).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 155435
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.