Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBM4 Mouse anti-Human, Clone: 3H3F12, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | RBM4 |
---|---|
Clone | 3H3F12 |
Concentration | 1.45 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6387
![]() ![]() |
Proteintech Group Inc
602921IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
May play a role in alternative splice site selection during pre-mRNA processing.Specifications
RBM4 | |
1.45 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
hLark, LARK, Lark homolog, RBM4, RBM4A, RNA binding motif protein 4, RNA binding motif protein 4a, RNA binding protein 4, ZCCHC21, ZCRB3A | |
RBM4 | |
IgG2a | |
Protein A | |
Antibody |
3H3F12 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9BWF3 | |
5936 | |
RBM4 Fusion Protein Ag2190 | |
Primary | |
-20°C | |
RBM4 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title