Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM6 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB329650 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB329650 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB329650 Supplier Novus Biologicals Supplier No. NBP297851MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBM6 Polyclonal antibody specifically detects RBM6 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen RBM6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias DEF-3, DEF3FLJ39446,3G2, DKFZp686B0877, FLJ36517, FLJ42036, g16, HLC-11, Lung cancer antigen NY-LU-12, lung cancer protooncogene 11, NY-LU-12, Protein G16, RNA binding motif protein 6, RNA-binding motif protein 6, RNA-binding protein 6, RNA-binding protein DEF-3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human RBM6 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10180
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.