Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBM8A Antibody (PCRP-RBM8A-1B4), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB390559 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390559 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390559 Supplier Novus Biologicals Supplier No. NBP308800AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBM8A Monoclonal antibody specifically detects RBM8A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen RBM8A
Applications Western Blot, Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-RBM8A-1B4
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias Binder of OVCA1-1, BOV-1, BOV-1A, BOV-1B, BOV-1C, MDS014, RBM8BRBM8, ribonucleoprotein RBM8, Ribonucleoprotein RBM8A, RNA binding motif protein 8A, RNA binding motif protein 8B, RNA-binding motif protein 8A, RNA-binding protein 8A, RNA-binding protein Y14, Y14, ZNRP, ZRNP1
Host Species Mouse
Immunogen Recombinant full-length human RBM8A protein (Uniprot: Q9Y5S9)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 9939
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.